Learn More
Eisco™ Class A Volumetric Pipet, Borosilicate Glass
Eisco Labs Volumetric Pipet, Borosilicate glass, 0.030 tolerance, Class A Glass, Individually wrapped
Supplier:  Eisco™ CH0559G
 Afficher plus de versions de ce produit
Numéro de catalogue. S28003
Il en reste null
Découvrez plus d'offres spéciales
Description
Description
  • Volumetric pipette featuring color coding for easy volume identification.
  • Made of high-quality Borosilicate 3.3 glass.
  • Autoclavable, with superior chemical durability.
  • Low temperature gradient - can withstand high temperatures and thermal shocks.
  • Class A tolerance ±0.010 mL, as per DIN ISO 648 standard.
  • Calibrated for delivery.
  • Each pipette is individually wrapped to keep clean.
Spécifications
Spécifications
Autoclavable
Non-sterile
0.8 oz.
25 mL
Serological Pipet
Borosilicate Glass
Blue
Blue
55.88 cm
Reusable
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS