missing translation for 'onlineSavingsMsg'
Learn More
Learn More
cis-beta-Methylstyrene (stabilized with TBC) 98.0+%, TCI America™
Supplier: TCI America M117410ML
Spécifications
| cis-beta-Methylstyrene (stabilized with TBC) | |
| -60°C | |
| 174°C | |
| MFCD00065137 | |
| 3295 | |
| QROGIFZRVHSFLM-KXFIGUGUSA-N | |
| [(Z)-prop-1-enyl]benzene | |
| 252324 | |
| ≥98.0% (GC) |
| 766-90-5 | |
| Colorless | |
| C9H10 | |
| 10 mL | |
| cis-1-Phenyl-1-propene, cis-Propenylbenzene | |
| CC=CC1=CC=CC=C1 | |
| 118.179 | |
| 118.18 | |
| Liquid |
Identifiants chimiques
| 766-90-5 | |
| 118.179 | |
| QROGIFZRVHSFLM-KXFIGUGUSA-N | |
| 252324 | |
| CC=CC1=CC=CC=C1 |
| C9H10 | |
| MFCD00065137 | |
| cis-1-Phenyl-1-propene, cis-Propenylbenzene | |
| [(Z)-prop-1-enyl]benzene |
Sécurité et manipulation
missing translation for 'einecsNumber' : (3)-0008
missing translation for 'tsca' : No