missing translation for 'onlineSavingsMsg'
Learn More
Learn More
cis-1,2-Cyclohexanedicarboxylic Anhydride 99.0+%, TCI America™
Supplier: TCI America C0478500G
Spécifications
| cis-1,2-Cyclohexanedicarboxylic Anhydride | |
| White | |
| C8H10O3 | |
| 500 g | |
| MUTGBJKUEZFXGO-OLQVQODUSA-N | |
| (3aS,7aR)-3a,4,5,6,7,7a-hexahydro-2-benzofuran-1,3-dione | |
| 641660 | |
| ≥99.0% (GC) |
| 13149-00-3 | |
| 160°C | |
| MFCD00005926 | |
| cis-hexahydroisobenzofuran-1,3-dione, cis-1,2-cyclohexanedicarboxylic anhydride, cis-hexahydrophthalic anhydride, unii-b52z7tnv87, cis-cyclohexane-1,2-dicarboxylic anhydride, hexahydrophthalic anhydride, cis, 3ar,7as-hexahydro-2-benzofuran-1,3-dione, 1,3-isobenzofurandione, hexahydro-, 3ar,7as-rel, 3ar,7as-hexahydroisobenzofuran-1,3-dione, 1,3-isobenzofurandione, hexahydro-, cis | |
| C1CCC2C(C1)C(=O)OC2=O | |
| 154.165 | |
| 154.17 | |
| Crystalline Lumps |
Identifiants chimiques
| 13149-00-3 | |
| 154.165 | |
| MUTGBJKUEZFXGO-OLQVQODUSA-N | |
| 641660 | |
| C1CCC2C(C1)C(=O)OC2=O |
| C8H10O3 | |
| MFCD00005926 | |
| cis-hexahydroisobenzofuran-1,3-dione, cis-1,2-cyclohexanedicarboxylic anhydride, cis-hexahydrophthalic anhydride, unii-b52z7tnv87, cis-cyclohexane-1,2-dicarboxylic anhydride, hexahydrophthalic anhydride, cis, 3ar,7as-hexahydro-2-benzofuran-1,3-dione, 1,3-isobenzofurandione, hexahydro-, 3ar,7as-rel, 3ar,7as-hexahydroisobenzofuran-1,3-dione, 1,3-isobenzofurandione, hexahydro-, cis | |
| (3aS,7aR)-3a,4,5,6,7,7a-hexahydro-2-benzofuran-1,3-dione |
Sécurité et manipulation
missing translation for 'einecsNumber' : (3)-2416
missing translation for 'tsca' : No