missing translation for 'onlineSavingsMsg'
Learn More
Learn More
cis-1,2-Cyclohexanedicarboxylic Anhydride 99.0+%, TCI America™
$33.56 - $134.21
Chemical Identifiers
| CAS | 13149-00-3 |
|---|---|
| Molecular Formula | C8H10O3 |
| Molecular Weight (g/mol) | 154.165 |
| MDL Number | MFCD00005926 |
| InChI Key | MUTGBJKUEZFXGO-OLQVQODUSA-N |
| Synonym | cis-hexahydroisobenzofuran-1,3-dione, cis-1,2-cyclohexanedicarboxylic anhydride, cis-hexahydrophthalic anhydride, unii-b52z7tnv87, cis-cyclohexane-1,2-dicarboxylic anhydride, hexahydrophthalic anhydride, cis, 3ar,7as-hexahydro-2-benzofuran-1,3-dione, 1,3-isobenzofurandione, hexahydro-, 3ar,7as-rel, 3ar,7as-hexahydroisobenzofuran-1,3-dione, 1,3-isobenzofurandione, hexahydro-, cis |
| PubChem CID | 641660 |
| IUPAC Name | (3aS,7aR)-3a,4,5,6,7,7a-hexahydro-2-benzofuran-1,3-dione |
| SMILES | C1CCC2C(C1)C(=O)OC2=O |
Chemical Identifiers
| 13149-00-3 | |
| 154.165 | |
| MUTGBJKUEZFXGO-OLQVQODUSA-N | |
| 641660 | |
| C1CCC2C(C1)C(=O)OC2=O |
| C8H10O3 | |
| MFCD00005926 | |
| cis-hexahydroisobenzofuran-1,3-dione, cis-1,2-cyclohexanedicarboxylic anhydride, cis-hexahydrophthalic anhydride, unii-b52z7tnv87, cis-cyclohexane-1,2-dicarboxylic anhydride, hexahydrophthalic anhydride, cis, 3ar,7as-hexahydro-2-benzofuran-1,3-dione, 1,3-isobenzofurandione, hexahydro-, 3ar,7as-rel, 3ar,7as-hexahydroisobenzofuran-1,3-dione, 1,3-isobenzofurandione, hexahydro-, cis | |
| (3aS,7aR)-3a,4,5,6,7,7a-hexahydro-2-benzofuran-1,3-dione |
Specifications
| 13149-00-3 | |
| 160°C | |
| MFCD00005926 | |
| cis-hexahydroisobenzofuran-1,3-dione, cis-1,2-cyclohexanedicarboxylic anhydride, cis-hexahydrophthalic anhydride, unii-b52z7tnv87, cis-cyclohexane-1,2-dicarboxylic anhydride, hexahydrophthalic anhydride, cis, 3ar,7as-hexahydro-2-benzofuran-1,3-dione, 1,3-isobenzofurandione, hexahydro-, 3ar,7as-rel, 3ar,7as-hexahydroisobenzofuran-1,3-dione, 1,3-isobenzofurandione, hexahydro-, cis | |
| C1CCC2C(C1)C(=O)OC2=O | |
| 154.165 | |
| 154.17 | |
| Crystalline Lumps |
| White | |
| C8H10O3 | |
| 25 g | |
| MUTGBJKUEZFXGO-OLQVQODUSA-N | |
| (3aS,7aR)-3a,4,5,6,7,7a-hexahydro-2-benzofuran-1,3-dione | |
| 641660 | |
| ≥99.0% (GC) | |
| cis-1,2-Cyclohexanedicarboxylic Anhydride |
Safety and Handling
EINECSNumber : (3)-2416
TSCA : No