Learn More
Chromium silicon monoxide, 99.85+% (metals basis), Thermo Scientific™
For vacuum deposition
Supplier: Thermo Scientific Chemicals 3637922
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Spécifications
| Chromium silicon monoxide | |
| Sintered Pieces | |
| (metals basis) | |
| chromium silicon monoxide, d-arg1,d-trp5 inverted exclamation mark currency7 inverted exclamation mark currency9,leu11-substance p | |
| ≥99.85% |
| 70320-14-8 | |
| 100 g | |
| MFCD02091702 | |
| Insoluble in water. |
Identifiants chimiques
| 70320-14-8 | |
| MFCD02091702 | |
| chromium silicon monoxide, d-arg1,d-trp5 inverted exclamation mark currency7 inverted exclamation mark currency9,leu11-substance p |
Sécurité et manipulation
GHS H Statement
H334-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.
P261-P272-P280g-P285-P302+P352-P304+P341-P333+P313-P342+P311-P363-P501c
H317-H334
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures