missing translation for 'onlineSavingsMsg'
Learn More

Chromium silicon monoxide, 99.85+% (metals basis), Thermo Scientific™

For vacuum deposition

$210.07 - $665.52

Chemical Identifiers

CAS 70320-14-8
MDL Number MFCD02091702
Synonym chromium silicon monoxide, d-arg1,d-trp5 inverted exclamation mark currency7 inverted exclamation mark currency9,leu11-substance p
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA3637914
View Documents
Thermo Scientific Chemicals
3637914
25 g
Each for $210.07
Only null left
 
AA3637922
View Documents
Thermo Scientific Chemicals
3637922
100 g
Each for $665.52
Only null left
 
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

70320-14-8
MFCD02091702
chromium silicon monoxide, d-arg1,d-trp5 inverted exclamation mark currency7 inverted exclamation mark currency9,leu11-substance p

Specifications

70320-14-8
25 g
chromium silicon monoxide, d-arg1,d-trp5 inverted exclamation mark currency7 inverted exclamation mark currency9,leu11-substance p
≥99.85%
(metals basis)
Sintered Pieces
MFCD02091702
Insoluble in water.
Chromium silicon monoxide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H334-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.

P261-P272-P280g-P285-P302+P352-P304+P341-P333+P313-P342+P311-P363-P501c

H317-H334

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'