missing translation for 'onlineSavingsMsg'
Learn More

Chromium AA Standard, Certified, 1000ppm ±5ppm (1mL = 1mg Cr), LabChem™

Supplier:  LabChem LC131201

 Afficher plus de versions de ce produit

Numéro de catalogue. LC131201


Il en reste null
Découvrez plus d'offres spéciales

Description

Description

Spécifications

Spécifications

Chromium AA Standard
Poly Bottle
99.72,0.28
Liquid
1000 ppm ±5 ppm (1mL = 1mg Cr)
Orange
KMUONIBRACKNSN-UHFFFAOYSA-N
dipotassium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium
500 mL
CHEBI:53444
Certified
1000ppm (1mL = 1mg Cr)
7778-50-9,7732-18-5
Passes Test
Cr2K2O7
K2Cr2O7
Soluble in water
[O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-].[K+].[K+]
294.182
24502
294.18

Identifiants chimiques

7778-50-9
294.182
24502
dipotassium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium
Cr2K2O7
KMUONIBRACKNSN-UHFFFAOYSA-N
CHEBI:53444
[O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-].[K+].[K+]
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
Sécurité et manipulation

Sécurité et manipulation

GHS H Statement
May cause an allergic skin reaction.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause genetic defects.
May cause cancer.
May damage fertility or the unborn child.
Harmful to aquatic life with long lasting effects.

GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Avoid breathing mist, vapors, spray.
Wear protective gloves, eye protection.
Wear respiratory protection.
Wash exposed skin thoroughly after handling.
Contaminated work clothing must not be allowed out of the workplace.
Avoid release to the environment.
If exposed or concerned: Get medical advice.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing and wash it before reuse.
If skin irritation or rash occurs: Get medical advice/attention.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
If experiencing respiratory symptoms: Call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.

Danger

Recommended Storage : Room Temperature

FDS
Documents

Documents

Promotions

Promotions