missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chlorothalonil, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Supplier: SPEX CertiPrep Group LLC S915
Comprend: Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
| Analyte | CAS No. | Concentration |
|---|---|---|
| Chlorothalonil | 1897-45-6 | 1000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Spécifications
| Chlorothalonil | |
| 1897-45-6,67-56-1 | |
| Certificate of Analysis reports target analytes to a ±2% accuracy and correct for raw material purity. | |
| For US EPA Method 8081 | |
| GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation | |
| CRQQGFGUEAVUIL-UHFFFAOYSA-N | |
| Methanol | |
| 265.902 | |
| 15910 |
| 2mL amber prescored glass ampule | |
| 0.1,99.9 | |
| 0.1,99.9 | |
| C8Cl4N2 | |
| Certified Reference Material | |
| C(#N)C1=C(C(=C(C(=C1Cl)Cl)Cl)C#N)Cl | |
| 2,4,5,6-tetrachlorobenzene-1,3-dicarbonitrile | |
| 1 mL | |
| CHEBI:3639 |
Identifiants chimiques
| 1897-45-6 | |
| 265.902 | |
| 15910 | |
| 2,4,5,6-tetrachlorobenzene-1,3-dicarbonitrile |
| C8Cl4N2 | |
| CRQQGFGUEAVUIL-UHFFFAOYSA-N | |
| CHEBI:3639 | |
| C(#N)C1=C(C(=C(C(=C1Cl)Cl)Cl)C#N)Cl |
Sécurité et manipulation
DOTInformation : Nonhazardous
Recommended Storage : Ambient
Product intended for laboratory use only