Learn More
Thermo Scientific™ CaptureSelect™ AAVX Ligand Leakage ELISA
Thermo Scientific™ CaptureSelect™ AAVX Ligand Leakage ELISA is designed to detect possible leached ligand of adeno-associated viruses (AAV) purified using POROS CaptureSelect AAVX Affinity Matrix.
$4148.67 - $4148.67
Spécifications
Product Line CaptureSelect™
Shipping Condition Dry Ice
voir les spécifications
Produits 1
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantité  
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantité  
81-035-2201
Afficher les documents
Thermo Scientific™
810352201
1 Assay
chaque for $4,148.67
Il en reste null
Description
Description
CaptureSelect AAVX Ligand Leakage ELISA is designed to detect possible leached ligand of adeno-associated viruses (AAV) purified using POROS CaptureSelect AAVX Affinity Matrix. The ligand, a 13-kD recombinant protein, is covalently coupled for high chemical stability and low leakage. Even when covalently attached, small amounts of the affinity ligand can leach off of the chromatography support and co-elute with the purified AAV.
This ELISA provides typical sensitivity and accurate quantitation of possible leached ligand greater than or equal to 1 ng/mL. POROS CaptureSelect AAVX Ligand Leakage ELISA can be used as a tool to aid in optimal purification process development and in routine quality control of in-process streams as well as final product.
Spécifications
Spécifications
CaptureSelect™
Dry Ice
Videos
FDS
missing translation for 'documents'
missing translation for 'documents'