missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ C13orf15 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5149916
Description
C13orf15 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
Modulates the activity of cell cycle-specific kinases. Enhances CDK1 activity. May contribute to the regulation of the cell cycle. May inhibit growth of glioma cells by promoting arrest of mitotic progression at the G2/M transition. Fibrogenic factor contributing to the pathogenesis of renal fibrosis through fibroblast activation.Spécifications
| C13orf15 | |
| Polyclonal | |
| Unconjugated | |
| RGCC | |
| 1190002H23Rik; bA157L14.2; C13orf15; regulator of cell cycle; regulator of cell cycle RGCC; Response gene to complement 32 protein; Rgc32; RGC-32; Rgcc | |
| Rabbit | |
| Protein A | |
| RUO | |
| 100625944, 117183, 28984, 66214 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q9DBX1, Q9H4X1, Q9Z2P4 | |
| RGCC | |
| Recombinant Response Gene To Complement 32 (RGC32) (Met1-Met137). | |
| 200 μL | |
| Primary | |
| Human, Mouse, Rat, Pig | |
| Antibody |