missing translation for 'onlineSavingsMsg'
Learn More

Buffer Solution pH 3.0 at 25°C, Certified, LabChem™

Supplier:  LabChem LC122502

 Afficher plus de versions de ce produit

Numéro de catalogue. LC122502


Il en reste null

Description

Description

Spécifications

Spécifications

Buffer Solution pH 3.0 at 25°C
Colorless
Formaldehyde; Carbon monoxide; Carbon dioxide
3.00 ±0.01
1 L
KHC8H4O4
1g/mL
Soluble in water
Certified
98.785,1.02,0.15,0.04,0.005
877-24-7 , 7647-01-0 , 50-00-0 , 67-56-1
1g/mL
Liquid
Passes Test
Poly Bottle
Traceable to NIST
204.23
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
FDS
Documents

Documents

Promotions

Promotions