Learn More
BTX™ BTXPress™ Cytofusion Medium C
BTXPress™ Cytofusion Medium C is designed to promote efficiency while preserving high cell viability during cell fusion.
Supplier: BTX™ BTX470001
Description
An advanced electrofusion buffer designed especially for hybridoma production. The low conductivity buffer is specially formulated to minimize cell turbulence during cell alignment and heating during electrofusion, for robust cell fusion efficiency and high cell viability. It is sterile-filtered from the highest quality non-animal, medical-grade reagents. It is the buffer of choice for many commercial biotech and pharmaceutical companies in their hybridoma generation process for monoclonal antibody discovery.
- Optimized for eukaryotic electrofusion applications
- Maximum fusion efficiency, high cell viability
- Stable environment for cell alignment
- Low conductivity for minimal heating
- Physiological pH and balanced osmolarity
- Contains no animal products
Spécifications
| Electrofusion Buffer | |
| Clear | |
| Non-animal | |
| None (Promoterless) | |
| BTX&trade ECM2001, Legacy ECM2001 | |
| 500 mL Buffer | |
| BTX Electroporation/Electrofusion | |
| 500 mL | |
| Sterile | |
| Liquid |
| 2°C to 8°C | |
| BTXpress Cytofusion Medium C | |
| Cells | |
| Electroporation | |
| Cell Fusion, Hybridoma Production | |
| 7.2 ± 0.2 | |
| Electrofusion Buffer | |
| Medical-grade | |
| 500 mL |