missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bromocresol Green - Methyl Red, Aqueous, for Alkalinity, Certified, LabChem™
Supplier: LabChem LC118707
Spécifications
| Bromocresol Green - Methyl Red | |
| 99.86,0.04,0.1 | |
| C21H14Br4O5S·Na | |
| Passes Test | |
| Certified | |
| 1g/mL | |
| 1g/mL | |
| 125 mL |
| Aqueous, for Alkalinity | |
| 845-10-3,62625-32-5 | |
| Soluble in water | |
| 720.06 | |
| White Poly | |
| Carbon monoxide; Carbon dioxide; Nitrogen oxides; Sulfur compounds | |
| Blue to Blue/Green | |
| Liquid |