missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bromocresol Green - Methyl Red, Aqueous, for Alkalinity, Certified, LabChem™
$47.07 - $490.73
Specifications
| Chemical Name or Material | Bromocresol Green - Methyl Red |
|---|---|
| Name Note | Aqueous, for Alkalinity |
| CAS Max % | 99.86,0.04,0.1 |
| CAS | 845-10-3,62625-32-5 |
| Linear Formula | C21H14Br4O5S·Na |
Specifications
| Bromocresol Green - Methyl Red | |
| 99.86,0.04,0.1 | |
| C21H14Br4O5S·Na | |
| Passes Test | |
| Certified | |
| Carbon monoxide; Carbon dioxide; Nitrogen oxides; Sulfur compounds | |
| Blue to Blue/Green |
| Aqueous, for Alkalinity | |
| 845-10-3,62625-32-5 | |
| Soluble in water | |
| 720.06 | |
| 1g/mL | |
| 1g/mL | |
| Liquid |