missing translation for 'onlineSavingsMsg'
Learn More

Bromocresol Green - Methyl Red, Aqueous, for Alkalinity, Certified, LabChem™

Supplier:  LabChem LC118701

 Afficher plus de versions de ce produit

Numéro de catalogue. LC118701


Il en reste null
Découvrez plus d'offres spéciales

Description

Description

Spécifications

Spécifications

Bromocresol Green - Methyl Red
99.86,0.04,0.1
C21H14Br4O5S·Na
Passes Test
Certified
1g/mL
1g/mL
500 mL
Aqueous, for Alkalinity
845-10-3,62625-32-5
Soluble in water
720.06
Amber Poly
Carbon monoxide; Carbon dioxide; Nitrogen oxides; Sulfur compounds
Blue to Blue/Green
Liquid
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
FDS
Documents

Documents

Promotions

Promotions