Learn More
alpha-Bromo-p-toluic acid, 97%
CAS: 6232-88-8 | C8H7BrO2 | 215.05 g/mol
Supplier: Thermo Scientific Chemicals 170400500
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spécifications
| α-Bromo-p-toluic acid | |
| 6232-88-8 | |
| 100.0 | |
| White to Beige | |
| 97% | |
| C8H7BrO2 | |
| MFCD00002567 | |
| 09,I,195 | |
| Solubility in water: insoluble. | |
| C1=CC(=CC=C1CBr)C(=O)O | |
| 215.05 | |
| 215.05 | |
| Crystalline Powder |
| 97% | |
| 96.0 | |
| 223.0°C to 227.0°C | |
| Authentic | |
| Glass bottle | |
| BrCH2C6H4CO2H | |
| 50 g | |
| 4-bromomethyl benzoic acid, alpha-bromo-p-toluic acid, benzoic acid, 4-bromomethyl, p-bromomethyl benzoic acid, 4-carboxybenzyl bromide, 4-bromomethyl-benzoic acid, a-bromo-p-toluic acid, p-carboxybenzyl bromide, p-bromomethylbenzoic acid | |
| CQQSQBRPAJSTFB-UHFFFAOYSA-N | |
| 4-(bromomethyl)benzoic acid | |
| 22599 | |
| 97% |
Identifiants chimiques
| 6232-88-8 | |
| 215.05 | |
| CQQSQBRPAJSTFB-UHFFFAOYSA-N | |
| 22599 | |
| C1=CC(=CC=C1CBr)C(=O)O |
| C8H7BrO2 | |
| MFCD00002567 | |
| 4-bromomethyl benzoic acid, alpha-bromo-p-toluic acid, benzoic acid, 4-bromomethyl, p-bromomethyl benzoic acid, 4-carboxybenzyl bromide, 4-bromomethyl-benzoic acid, a-bromo-p-toluic acid, p-carboxybenzyl bromide, p-bromomethylbenzoic acid | |
| 4-(bromomethyl)benzoic acid |
Sécurité et manipulation
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 228-343-3
missing translation for 'tsca' : TSCA
RUO – Research Use Only