missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Ball Valve Lockout Devices

Fit all major brand ball valves

$35.94 - $72.56

Spécifications

Product Type Ball Valve Lockout
Color Red
Certifications/Compliance OSHA
For Use With (Application) To help prevent accidental start-ups
voir les spécifications

Produits 4
Numéro de catalogue Numéro du manufacturier. For Use With (Equipment) Diameter (English) Material Prix Quantity & Availability  
Numéro de catalogue Numéro du manufacturier. For Use With (Equipment) Diameter (English) Material Prix Quantity & Availability  
17987275D
Afficher les documents Détails de la promotion
Brady™
65666
Valve Size: 0.25 to 1 in. 0.31 in. Nylon
chaque for $35.94
Il en reste null
 
17987275E
Afficher les documents Détails de la promotion
Brady™
65669
Valve Size: 1.25 to 3 in. 0.31 in. Powder-coated Steel
chaque for $71.25
Il en reste null
 
19016530
Afficher les documents Détails de la promotion
Brady™
65693
Valve Size: 1.25 to 3 in. 0.35 in. Powder-coated Steel
chaque for $72.56
Il en reste null
 
19016529
Afficher les documents Détails de la promotion
Brady™
65692
Valve Size: 0.25 to 1 in. 0.35 in. Powder-coated Steel
chaque for $41.17
Il en reste null
 
Description

Description

  • Lock ball valves to help prevent accidental start-ups
  • Patented, easy-to-use one-piece design

Compliance

Assist in compliance with OSHA's lockout standard.

Spécifications

Spécifications

Ball Valve Lockout
Red
OSHA
To help prevent accidental start-ups
Videos
FDS
Documents

Documents