Learn More
Fisherbrand™ Borosilicate Cylinders with Pouring Lip
Heavy-wall construction resists breakage
 Afficher plus de versions de ce produit
Numéro de catalogue. 08-530K
Il en reste null
Description
Description
Heavy-wall construction resists breakage
  • Ungraduated
  • Fused round base
  • For use with hydrometers
Spécifications
Spécifications
Round
Ungraduated Cylinder
14.17 in.
41 mm
Borosilicate Glass
Round
375 mm
1.61 in.
350 mL
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS