missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ BIOshell™ A160 Peptide CN HPLC Column, 160Å, 4.7 μm Particle Size

Numéro de catalogue. 111008371
Change view
Click to view available options
Length (Metric):
3 cm
5 cm
7.5 cm
10 cm
15 cm
Diameter (Metric):
2.1 mm
3 mm
4.6 mm
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Length (Metric) Diameter (Metric)
111008371 3 cm 2.1 mm
111007929 5 cm 2.1 mm
111007670 7.5 cm 2.1 mm
111007515 10 cm 2.1 mm
111007060 15 cm 2.1 mm
111008372 3 cm 3 mm
111007930 5 cm 3 mm
111007516 10 cm 3 mm
111007061 15 cm 3 mm
111007931 5 cm 4.6 mm
111007517 10 cm 4.6 mm
111007062 15 cm 4.6 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Numéro de catalogue. 111008371 Fournisseur MilliporeSigma™ Supelco™ Code fournisseur 67061U
Il en reste null

BIOshell™ 160 Å Peptide CN columns provide an alternative selectivity to the traditional C18 bonded phases.

The introduction of the cyano group enables peptides and small proteins to be separated due to a combination of hydrophobicity and dipole-dipole interactions making this column an excellent choice for biomolecules that are not resolved on traditional C18 columns. These columns contain advanced Fused-Core particles with pores strategically sized to 160 Å to enable optimized peptide diffusion. This attribute makes these columns an excellent choice for peptide mapping. Additionally, the sterically-protected cyanopropyl ligands provide extra stability allowing the columns to be used at an extended pH range (2-9) and high temperatures (up to 90°C).

Spécifications

Diameter (Metric) 2.1 mm
Length (Metric) 3 cm
Particle Size 4.7 μm
pH 1 to 8
Pore Size 160 Å
Surface Area 60 m2/g
Stationary Phase Cyano
Product Line BIOshell
Matrix Cyano Phase/Spherical Silica Particle Platform
Carbon Load 1.5%
Endcapped Yes
Phase Reversed Phase
Material Stainless Steel
Technique Fused-Core HPLC
Afficher plus de résultats Afficher moins de résultats

BIOshell is a trademark of Sigma-Aldrich Co. LLC