missing translation for 'onlineSavingsMsg'
Learn More

Bexarotene, 98%, Thermo Scientific Chemicals

Synthetic retinoid analog with specific affinity for the retinoid X receptor

Supplier:  Thermo Scientific Chemicals J63701MC

 Afficher plus de versions de ce produit

Numéro de catalogue. AAJ63701MC


Il en reste null
Découvrez plus d'offres spéciales

Description

Description

Bexarotene is used as a Synthetic retinoid analog with specific affinity for the retinoid X receptor, an antineoplastic agent, already approved as an oral antineoplastic agent for cutaneous T cell lymphoma and being investigated against other cancers. A study has found that bexarotene in a mouse Alzheimer's model lowered the most toxic form of β-amyloid peptide and increased cognitive ability.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bexarotene is used as a Synthetic retinoid analog with specific affinity for the retinoid X receptor, an antineoplastic agent, already approved as an oral antineoplastic agent for cutaneous T cell lymphoma and being investigated against other cancers. A study has found that bexarotene in a mouse Alzheimer′s model lowered the most toxic form of β-amyloid peptide and increased cognitive ability.

Solubility
Soluble in DMSO and ethanol

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
Spécifications

Spécifications

Bexarotene
220°C to 225°C
C24H28O2
100 mg
bexarotene, targretin, targrexin, targretyn, targret, bexarotenum, bexarotene usan, bexaroteno, bexaroten, targretin-gel
NAVMQTYZDKMPEU-UHFFFAOYSA-N
4-[1-(3,5,5,8,8-pentamethyl-5,6,7,8-tetrahydronaphthalen-2-yl)ethenyl]benzoic acid
82146
348.48
Powder
153559-49-0
White
MFCD00932428
14,1194
Soluble in DMSO and ethanol
CC1=CC2=C(C=C1C(=C)C1=CC=C(C=C1)C(O)=O)C(C)(C)CCC2(C)C
348.49
CHEBI:50859
≥99%

Identifiants chimiques

153559-49-0
348.49
NAVMQTYZDKMPEU-UHFFFAOYSA-N
82146
4-[1-(3,5,5,8,8-pentamethyl-5,6,7,8-tetrahydronaphthalen-2-yl)ethenyl]benzoic acid
C24H28O2
MFCD00932428
bexarotene, targretin, targrexin, targretyn, targret, bexarotenum, bexarotene usan, bexaroteno, bexaroten, targretin-gel
CHEBI:50859
CC1=CC2=C(C=C1C(=C)C1=CC=C(C=C1)C(O)=O)C(C)(C)CCC2(C)C
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
Sécurité et manipulation

Sécurité et manipulation

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

missing translation for 'rtecsNumber' : DH6834830

missing translation for 'tsca' : No

Recommended Storage : Store at -20°C

FDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only