missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BCCIP Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$344.88 - $344.88
Spécifications
| Antigen | BCCIP |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Liquid |
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantité | |||||
|---|---|---|---|---|---|---|---|---|---|
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantité | |||||
501562019
|
Bethyl Laboratories Inc
A302196AT |
20 μL |
chaque for $344.88
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
Description
The recommended shelf life for this product is 1 year from date of receipt.
May promote cell cycle arrest by enhancing the inhibition of CDK2 activity by CDKN1A. May be required for repair of DNA damage by homologous recombination in conjunction with BRCA2. May not be involved in non-homologous end joining (NHEJ).Spécifications
| BCCIP | |
| Polyclonal | |
| Liquid | |
| RUO | |
| TBS with 0.1% BSA and 0.09% sodium azide | |
| TOK-1, TOK1, BCCIPalpha, BCCIPbeta, BRCA2 and CDKN1A-interacting protein, BRCA2 and Cip1/p21 interacting protein, TOK-1alpha, TOK-1beta, cdk inhibitor p21 binding protein, p21- and CDK-associated protein 1, protein TOK-1 | |
| BCCIP | |
| IgG | |
| Antigen affinity chromatography | |
| Antibody |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q9P287 | |
| 56647 | |
| Between 1 and 50 | |
| Primary | |
| 4° C | |
| BCCIP |