Learn More
Eisco™ Class B Glass Volumetric Flask with Glass Stopper
Class B Glass Volumetric Flask with Glass Stopper features a single graduation and blue markings.
Supplier:  Eisco™ CH0442G
 Afficher plus de versions de ce produit
Numéro de catalogue. S23816
Il en reste null
Découvrez plus d'offres spéciales
Description
Description
  • Flat bottom design
  • Single graduation mark
  • Includes fitted glass stopper
  • Blue printed markings
  • Class B, in compliance with ASTM-E288
  • Autoclavable
Warranty
1 year limited manufacturer warranty.
Spécifications
Spécifications
33.8 oz.
Clear
Clear
1000 mL
1
Volumetric Flask
±0.6 mL
Reusable
Beaded
B
Glass Stopper
Stopper No. 22
ASTM-E288
Borosilicate Glass 3.3
Volumetric flask
Borosilicate Glass
Long
Straight-sided, Round
Autoclavable
Yes
Non-sterile
Flat
Non-treated
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS