missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARID3B Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories Inc A302564AT
Description
The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections.
This gene encodes a member of the ARID family of DNA-binding proteins. The encoded protein is homologous with two proteins that bind to the retinoblastoma gene product, and also with the mouse Bright and Drosophila dead ringer proteins. A pseudogene on chromosome 1p31 exists for this gene. Members of the ARID family have roles in embryonic patterning, cell lineage gene regulation, cell cycle control, transcriptional regulation and possibly in chromatin structure modification.Spécifications
| ARID3B | |
| Polyclonal | |
| TBS with 0.1% BSA and 0.09% sodium azide | |
| Q8IVW6 | |
| ARID3B | |
| Between 100 and 150 | |
| 10 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunohistochemistry, Western Blot | |
| Unconjugated | |
| ARID3B | |
| ARID domain-containing protein 3B; Bright-like; Bright and dead ringer protein; BDP; DRIL2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10620 | |
| 4° C | |
| Liquid |