missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ TaqMan™ Fast Universal PCR Master Mix (2X), no AmpErase™ UNG
TaqMan™ Fast Universal PCR Master Mix contains all of the components, excluding the template and primers, for superior real-time qPCR.
$602.00 - $11,150.00
Specifications
| Concentration | 2X |
|---|---|
| Detection Method | Primer-probe |
| Format | Tube |
| GC-Rich PCR Performance | High |
| PCR Method | qPCR |
Requires: Template and primers
| Catalog Number | Mfr. No. | No. of Reactions | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | No. of Reactions | Price | Quantity & Availability | |||||
4366072
|
Applied Biosystems™
4366072 |
500 Reactions |
Each for $1,248.00
|
|
|||||
4367846
|
Applied Biosystems™
4367846 |
5000 Reactions |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
4352042
|
Applied Biosystems™
4352042 |
250 Reactions |
Each for $602.00
|
|
|||||
4366073
|
Applied Biosystems™
4366073 |
1250 Reactions |
Each for $2,895.00
|
|
|||||
4364103
|
Applied Biosystems™
4364103 |
2500 Reactions |
Each for $5,540.00
|
|
|||||
Description
- Results in 40 minutes for 40 cycles of PCR in a 20 μL reaction volume when using the Applied Biosystems 7900HT and Fast 7500 Fast real-time PCR systems
- The optimized formulation contains AmpliTaq Fast DNA Polymerase, UP, a highly purified DNA polymerase designed to allow instant hot start, minimizing non-specific product formation and allowing room temperature reaction setup
- A proprietary ROX dye serves as a passive internal reference to normalize non-PCR-related fluorescence fluctuations, for superb precision on Applied Biosystems real-time PCR instruments
- Contains everything needed for TaqMan-based qPCR amplification and detection in a single-tube format
- TaqMan Fast Universal PCR Master Mix contains all of the components, excluding the template and primers, for superior real-time qPCR
- Compatible with TaqMan Gene Expression assays
- Store at 2°C to 8°C
Order Info
Guaranteed minimum shelf life is 60 days.
Specifications
| 2X | |
| Tube | |
| qPCR | |
| Fast | |
| ROX (Pre-mixed) | |
| Fast Universal PCR Master Mix | |
| 2 X |
| Primer-probe | |
| High | |
| AmpliTaq DNA Polymerase | |
| 7500 Fast System, 7900HT Fast System, StepOne™, Fast Mode, StepOnePlus™, Fast Mode | |
| AmpErase, TaqMan | |
| Gene Expression, Genotyping, miRNA analysis | |
| DNA (Genomic), cDNA |
For Research Use Only. Not for use in diagnostic procedures.