missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ TaqMan™ Fast Universal PCR Master Mix (2X), no AmpErase™ UNG

Catalog No. 4364103
Encompass
Change view
Click to view available options
No. of Reactions:
1250 Reactions
250 Reactions
2500 Reactions
500 Reactions
5000 Reactions
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
4364103 2500 Reactions
4366072 500 Reactions
4367846 5000 Reactions
4352042 250 Reactions
4366073 1250 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 4364103 Supplier Applied Biosystems™ Supplier No. 4364103
Only null left

Requires

Template and primers

TaqMan™ Fast Universal PCR Master Mix contains all of the components, excluding the template and primers, for superior real-time qPCR.

  • Results in 40 minutes for 40 cycles of PCR in a 20 μL reaction volume when using the Applied Biosystems 7900HT and Fast 7500 Fast real-time PCR systems
  • The optimized formulation contains AmpliTaq Fast DNA Polymerase, UP, a highly purified DNA polymerase designed to allow instant hot start, minimizing non-specific product formation and allowing room temperature reaction setup
  • A proprietary ROX dye serves as a passive internal reference to normalize non-PCR-related fluorescence fluctuations, for superb precision on Applied Biosystems real-time PCR instruments
  • Contains everything needed for TaqMan-based qPCR amplification and detection in a single-tube format
  • TaqMan Fast Universal PCR Master Mix contains all of the components, excluding the template and primers, for superior real-time qPCR
  • Compatible with TaqMan Gene Expression assays
  • Store at 2°C to 8°C

Order Info

Guaranteed minimum shelf life is 60 days.

Specifications

Concentration 2X
Content And Storage TaqMan™ Fast Universal PCR Master Mix, No AmpErase™ UNG Kit contains 10 packs of 2 tubes (1.25 ml each), sufficient for 2500 PCR reactions (20 μl each).

Store at 2°C to 8°C.

Guaranteed minimum shelf life is 60 days (exact expiry date printed on product and CofA).

Detection Method Primer-probe
Format Tube
GC-Rich PCR Performance High
PCR Method qPCR
Polymerase AmpliTaq DNA Polymerase
Reaction Speed Fast
For Use With (Equipment) 7500 Fast System, 7900HT Fast System, StepOne™, Fast Mode, StepOnePlus™, Fast Mode
Passive Reference Dye ROX (Pre-mixed)
Product Line AmpErase, TaqMan
Product Type Fast Universal PCR Master Mix
Quantity 2,500 reactions
Sufficient For 2500 Reactions
For Use With (Application) Gene Expression, Genotyping, miRNA analysis
Fidelity (vs. Taq) 2 X
No. of Reactions 2500 Reactions
Sample Type DNA (Genomic), cDNA
Volume 20 μL
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.