missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ TaqMan™ Fast Universal PCR Master Mix (2X), no AmpErase™ UNG

TaqMan™ Fast Universal PCR Master Mix contains all of the components, excluding the template and primers, for superior real-time qPCR.

Supplier:  Applied Biosystems™ 4352042

Encompass

 View more versions of this product

Catalog No. 4352042


Get Up to $3,000 of qPCR Products with Purchase
View all promotions
Only null left

Requires : Template and primers

Description

Description

  • Results in 40 minutes for 40 cycles of PCR in a 20 μL reaction volume when using the Applied Biosystems 7900HT and Fast 7500 Fast real-time PCR systems
  • The optimized formulation contains AmpliTaq Fast DNA Polymerase, UP, a highly purified DNA polymerase designed to allow instant hot start, minimizing non-specific product formation and allowing room temperature reaction setup
  • A proprietary ROX dye serves as a passive internal reference to normalize non-PCR-related fluorescence fluctuations, for superb precision on Applied Biosystems real-time PCR instruments
  • Contains everything needed for TaqMan-based qPCR amplification and detection in a single-tube format
  • TaqMan Fast Universal PCR Master Mix contains all of the components, excluding the template and primers, for superior real-time qPCR
  • Compatible with TaqMan Gene Expression assays
  • Store at 2°C to 8°C

Order Info

Guaranteed minimum shelf life is 60 days.

Specifications

Chemical Identifiers

2X
Tube
qPCR
Fast
ROX (Pre-mixed)
Fast Universal PCR Master Mix
2 X
Primer-probe
High
AmpliTaq DNA Polymerase
7500 Fast System, 7900HT Fast System, StepOne™, Fast Mode, StepOnePlus™, Fast Mode
AmpErase, TaqMan
Gene Expression, Genotyping, miRNA analysis
DNA (Genomic), cDNA

Specifications

2X
Primer-probe
High
AmpliTaq DNA Polymerase
7500 Fast System, 7900HT Fast System, StepOne™, Fast Mode, StepOnePlus™, Fast Mode
AmpErase, TaqMan
250 reactions
250 Reactions
2 X
DNA (Genomic), cDNA
TaqMan™ Fast Universal PCR Master Mix, No AmpErase™ UNG Kit contains two tubes of 1.25 mL each sufficient for 250 20-μL PCR reactions. Store at 2-8°C

Guaranteed minimum shelf life is 60 days (exact expiry date printed on product and CofA).
Tube
qPCR
Fast
ROX (Pre-mixed)
Fast Universal PCR Master Mix
Wet Ice
Gene Expression, Genotyping, miRNA analysis
250 Reactions
20 μL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.