missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ TaqMan™ Fast Universal PCR Master Mix (2X), no AmpErase™ UNG
TaqMan™ Fast Universal PCR Master Mix contains all of the components, excluding the template and primers, for superior real-time qPCR.
Supplier: Applied Biosystems™ 4366073
Requires : Template and primers
Description
- Results in 40 minutes for 40 cycles of PCR in a 20 μL reaction volume when using the Applied Biosystems 7900HT and Fast 7500 Fast real-time PCR systems
- The optimized formulation contains AmpliTaq Fast DNA Polymerase, UP, a highly purified DNA polymerase designed to allow instant hot start, minimizing non-specific product formation and allowing room temperature reaction setup
- A proprietary ROX dye serves as a passive internal reference to normalize non-PCR-related fluorescence fluctuations, for superb precision on Applied Biosystems real-time PCR instruments
- Contains everything needed for TaqMan-based qPCR amplification and detection in a single-tube format
- TaqMan Fast Universal PCR Master Mix contains all of the components, excluding the template and primers, for superior real-time qPCR
- Compatible with TaqMan Gene Expression assays
- Store at 2°C to 8°C
Order Info
Guaranteed minimum shelf life is 60 days.
Chemical Identifiers
| 2X | |
| Tube | |
| qPCR | |
| Fast | |
| ROX (Pre-mixed) | |
| Fast Universal PCR Master Mix | |
| 2 X |
| Primer-probe | |
| High | |
| AmpliTaq DNA Polymerase | |
| 7500 Fast System, 7900HT Fast System, StepOne™, Fast Mode, StepOnePlus™, Fast Mode | |
| AmpErase, TaqMan | |
| Gene Expression, Genotyping, miRNA analysis | |
| DNA (Genomic), cDNA |
Specifications
| 2X | |
| Primer-probe | |
| High | |
| AmpliTaq DNA Polymerase | |
| 7500 Fast System, 7900HT Fast System, StepOne™, Fast Mode, StepOnePlus™, Fast Mode | |
| AmpErase, TaqMan | |
| 1,250 reactions | |
| Gene Expression, Genotyping, miRNA analysis | |
| 1250 Reactions | |
| 1.25 mL |
| This 5-pack contains five kits, each containing 2 tubes (1.25 ml each) of 2X TaqMan™ Fast Universal PCR Master Mix. Store at 2°C to 8°C. Guaranteed minimum shelf life is 60 days (exact expiry date printed on product and CofA). |
|
| Tube | |
| qPCR | |
| Fast | |
| ROX (Pre-mixed) | |
| Fast Universal PCR Master Mix | |
| 250 Reactions | |
| 2 X | |
| DNA (Genomic), cDNA |
For Research Use Only. Not for use in diagnostic procedures.