missing translation for 'onlineSavingsMsg'
Learn More
Learn More
tetratricopeptide repeat domain 1, Mouse, Clone: 4E3, Abnova™
Mouse monoclonal antibody raised against a partial recombinant TTC1.
Supplier: Abnova Corporation H00007265M01
Description
Sequence: LRRAELYEKTDKLDEALEDYKSILEKDPSIHQAREACMRLPKQIEERNERLKEEMLGKLKDLGNLVLRPFGLSTENFQIKQDSSTGSYSINFVQNPNNNRSpécifications
| tetratricopeptide repeat domain 1 | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_003314 | |
| TTC1 | |
| TTC1 (NP_003305, 193 a.a. ∼ 292 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Yes | |
| 7265 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2b κ |
| ELISA, Immunohistochemistry (PFA fixed), Western Blot | |
| 4E3 | |
| Mouse monoclonal antibody raised against a partial recombinant TTC1. | |
| TTC1 | |
| FLJ46404/TPR1 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| Antibody |