Learn More
RNF168 Rabbit anti-Human, Mouse, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier:  MilliporeSigma ABE367
Numéro de catalogue. ABE367MI
Il en reste null
Découvrez plus d'offres spéciales
Description
Description
Use Anti-RNF168 Antibody (Rabbit Polyclonal Antibody) validated in western blot, immunocytochemistry, immunofluorescence to detect RNF168 also known as E3 ubiquitin-protein ligase RNF168.
Spécifications
Spécifications
RNF168
Polyclonal
Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Rabbit
Affinity Purified
Primary
Human
Immunocytochemistry, Immunofluorescence, Western Blot
Unconjugated
RNF168
Histidine-tagged recombinant protein corresponding to the C-terminus of human RNF168.
100 μg
NP_689830
2-8°C for 12 months
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS