Learn More
Phospho-AMPK1 (Thr174) Rabbit anti-Human, Rat, Mouse, Canine, Bovine, Chicken, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma 09290
Description
Anti-phospho-AMPK1 (Thr174) Antibody detects level of phospho-AMPK1 (Thr174) and has been published and validated for use in western blot, immunohistochemistry.Spécifications
| phospho-AMPK1 (Thr174) | |
| Polyclonal | |
| Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH7.4) 150mM NaCl with 0.05% sodium azide. | |
| Rabbit | |
| Affinity Purified | |
| Primary | |
| Bovine, Canine, Chicken, Human, Mouse, Rat |
| Immunohistochemistry, Western Blot | |
| Unconjugated | |
| AMPK; AMPK1; AMPKa1; EC 2.7.11.1; MGC33776; MGC57364 | |
| KLH-conjugated linear peptide corresponding to phosphorylated human AMPK1 (Thr174). | |
| 100 μg | |
| NP_006242 | |
| 2-8°C for 12 months |
For Research Use Only