Learn More
Integrin α4 Rat anti-Human, Mouse, Clone: PS/2, MilliporeSigma™
Rat Monoclonal Antibody
Supplier:  MilliporeSigma CBL1304
Numéro de catalogue. CBL1304MI
Il en reste null
Découvrez plus d'offres spéciales
Description
Description
Anti-Integrin α4 Antibody, clone PS/2 detects level of Integrin α4 and has been published and validated for use in flow cytometry, immunofluoresence, immunoprecipitation, immunohistochemistry.
Spécifications
Spécifications
Integrin α4
Monoclonal
Unconjugated
NM_000885.4
Rat
Primary
2-8°C in undiluted aliquots for up to 12 months.
IgG2b κ
Flow Cytometry, Immunofluorescence, Immunohistochemistry, Immunoprecipitation
PS/2
Purified immunoglobulin in 100mM borate buffered saline containing no preservatives.
ITGA4, IA4, MGC90518, VLA-4, CD49D, CD49d
500 μg
Human, Mouse
Purified
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS