Learn More
FH, Mouse, Clone: 5D8, Abnova™
Mouse monoclonal antibody raised against a full-length recombinant FH.
Supplier: Abnova Corporation H00002271M02
Description
The protein encoded by this gene is an enzymatic component of the tricarboxylic acid (TCA) cycle, or Krebs cycle, and catalyzes the formation of L-malate from fumarate. It exists in both a cytosolic form and an N-terminal extended form, differing only in the translation start site used. The N-terminal extended form is targeted to the mitochondrion, where the removal of the extension generates the same form as in the cytoplasm. It is similar to some thermostable class II fumarases and functions as a homotetramer. Mutations in this gene can cause fumarase deficiency and lead to progressive encephalopathy. [provided by RefSeq
Sequence: VPSFWPPNAARMASQNSFRIEYDTFGELKVPNDKYYGAQTVRSTMNFKIGGVTERMPTPVIKAFGILKRAAAEVNQDYGLDPKIANAIMKAADEVAEGKLNDHFPLVVWQTGSGTQTNMNVNEVISNRAIEMLGGELGSKIPVHPNDHVNKSQSSNDTFPTAMHIAAAIEVHEVLLPGLQKLHDALDAKSKEFAQIIKIGRTHTQDAVPLTLGQEFSGYVQQVKYAMTRIKAAMPRIYELAAGGTAVGTGLNTRIGFAEKVAAKVAALTGLPFVTAPNKFEALAAHDALVELSGAMNTTACSLMKIANDIRFLGSGPRSGLGELILPENEPGSSIMPGKVNPTQCEAMTMVAAQVMGNHVAVTVGGSNGHFELNVFKPMMIKNVLHSARLLGDASVSFTENCVVGIQANTERINKLMNESLMLVTALNPHIGYDKAAKIAKTAHKNGSTLKETAIELGYLTAEQFDEWVKPKDMLGPKSpécifications
| FH | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| BC003108 | |
| FH | |
| FH (AAH03108, 33 a.a. ∼ 510 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b κ |
| ELISA, Western Blot | |
| 5D8 | |
| Mouse monoclonal antibody raised against a full-length recombinant FH. | |
| FH | |
| HLRCC/LRCC/MCL/MCUL1 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 2271 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |