Learn More
phenylalanyl-tRNA synthetase, alpha subunit, Mouse, Clone: 2D8, Abnova™
Mouse monoclonal antibody raised against a partial recombinant FARSLA.
Supplier: Abnova Corporation H00002193M01
Description
Aminoacyl-tRNA synthetases are a class of enzymes that charge tRNAs with their cognate amino acids. This gene encodes a product which is similar to the catalytic subunit of prokaryotic and Saccharomyces cerevisiae phenylalanyl-tRNA synthetases (PheRS). This gene product has been shown to be expressed in a tumor-selective and cell cycle stage- and differentiation-dependent manner, the first member of the tRNA synthetase gene family shown to exhibit this type of regulated expression [provided by RefSeq
Sequence: KVGFSKAMSNKWIRVDKSAADGPRVFRVVDSMEDEVQRRLQLVRGGQAEKLGEKERSELRKRKLLAEVTLKTYWVSKGSAFSTSISKQETELSPEMISSGSSpécifications
| phenylalanyl-tRNA synthetase, alpha subunit | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_004461 | |
| FARSA | |
| FARSLA (NP_004452, 101 a.a. ∼ 201 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Western Blot | |
| 2D8 | |
| Mouse monoclonal antibody raised against a partial recombinant FARSLA. | |
| FARSA | |
| CML33/FARSL/FARSLA/FRSA/PheHA | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| 2193 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG1 κ |