missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Apolipoprotein B Mouse anti-Human, Clone: B1, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MAB012
Description
This Anti-Apolipoprotein B Antibody, clone B1 is validated for use in ELISA, RIA for the detection of Apolipoprotein B.Spécifications
| Apolipoprotein B | |
| Monoclonal | |
| Unconjugated | |
| NM_000384.2 | |
| Mouse | |
| 500 μg | |
| Human | |
| Purified |
| ELISA, Radioimmune Assays (RIA) | |
| B1 | |
| Liquid, in 0.015Mpotassium phosphate buffer with 0.15M NaCl, 0.1% NaN3, pH 7.2. | |
| APOB, apoB-48, FLDB, apoB-100 | |
| Protein A purfied | |
| Primary | |
| −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles. | |
| IgG1 |
For Research Use Only