missing translation for 'onlineSavingsMsg'
Learn More

amelogenin (amelogenesis imperfecta 1, X-linked), Mouse, Clone: 3B5, Abnova™

Mouse monoclonal antibody raised against a partial recombinant AMELX.

Supplier:  Abnova Corporation H00000265M03

Numéro de catalogue. 89-000-517


Il en reste null
Découvrez plus d'offres spéciales

Description

Description

This gene encodes a member of the amelogenin family of extracellular matrix proteins. Amelogenins are involved in biomineralization during tooth enamel development. Mutations in this gene cause X-linked amelogenesis imperfecta. Alternative splicing results in multiple transcript variants encoding different isoforms. [provided by RefSeq

Sequence: PVIPQQPMMPVPGQHSMTPIQHHQPNLPPPAQQPYQPQPVQPQPHQPMQPQPPVHPMQPLPPQPPLPPMFPMQPLPPMLPDLTLEAWPSTDKTKREEVD
Spécifications

Spécifications

amelogenin (amelogenesis imperfecta 1, X-linked)
Monoclonal
Unconjugated
PBS with no preservative; pH 7.4
NM_001142
AMELX
AMELX (NP_001133, 93 a.a. ∼ 191 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
50 μg
Yes
265
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
IgG2b κ
ELISA, Western Blot
3B5
Mouse monoclonal antibody raised against a partial recombinant AMELX.
AMELX
AIH1/ALGN/AMG/AMGL/AMGX
Mouse
Affinity Purified
RUO
Primary
Human
Antibody
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
FDS
Documents

Documents

Promotions

Promotions