missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha-1-B glycoprotein, Mouse. Clone: 4F6, Abnova™
Mouse monoclonal antibody raised against a full-length recombinant A1BG
Supplier: Abnova Corporation H00000001M02
Description
The protein encoded by this gene is a plasma glycoprotein of unknown function. The protein shows sequence similarity to the variable regions of some immunoglobulin supergene family member proteins. [provided by RefSeq
Sequence: MAPVSWITPGLKTTAVCRGVLRGVTFLLRREGDHEFLEVPEAQEDVEATFPVHQPGNYSCSYRTDGEGALSEPSATVTIEELAAPPPPVLMHHGESSQVLHPGNKVTLTCVAPLSGVDFQLRRGEKELLVPRSSTSPDRIFFHLNAVALGDGGHYTCRYRLHDNQNGWSGDSAPVELILSDETLPAPEFSPEPESGRALRLRCLAPLEGARFALVREDRGGRRVHRFQSPAGTEALFELHNISVADSANYSCVYVDLKPPFGGSAPSERLELHVDGPPPRPQLRATWSGAVLAGRDAVLRCEGPIPDVTFELLREGETKAVKTVRTPGAAANLELIFVGPQHAGNYRCRYRSWVPHTFESELSDPVELLVAESSpécifications
| alpha-1-B glycoprotein | |
| Monoclonal | |
| 0.1mg/mL | |
| Mouse monoclonal antibody raised against a full length recombinant A1BG. | |
| A1BG | |
| A1B/ABG/DKFZp686F0970/GAB/HYST2477 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| 1 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
| ELISA | |
| 4F6 | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| BC035719 | |
| A1BG | |
| A1BG (AAH35719, 1 a.a. ∼ 373 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |