missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Hydroxide, 20% (v/v), Ricca Chemical

Supplier:  Ricca Chemical Company R063500020F

 Afficher plus de versions de ce produit

Numéro de catalogue. R063500020F


Il en reste null
Découvrez plus d'offres spéciales

Description

Description

Spécifications

Spécifications

Ammonium Hydroxide
1336-21-6,7732-18-5
MFCD00066650
ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh
VHUUQVKOLVNVRT-UHFFFAOYSA-N
amine hydrate
14923
19.3 to 20.7%
Cubitainer
Liquid
Colorless
H5NO
20% (v/v)
20 L
N.O
35.05
CHEBI:18219
Laboratory
Aqueous Solution

Identifiants chimiques

1336-21-6
35.05
VHUUQVKOLVNVRT-UHFFFAOYSA-N
14923
amine hydrate
H5NO
MFCD00066650
ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh
CHEBI:18219
N.O
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
FDS
Documents

Documents

Promotions

Promotions