missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Hydroxide, 20% (v/v), Ricca Chemical

$107.97 - $498.25

Chemical Identifiers

CAS 1336-21-6
Molecular Formula H5NO
Molecular Weight (g/mol) 35.05
MDL Number MFCD00066650
InChI Key VHUUQVKOLVNVRT-UHFFFAOYSA-N
Synonym ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh
PubChem CID 14923
ChEBI CHEBI:18219
IUPAC Name amine hydrate
SMILES N.O
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
635-16
SDS View Documents
Ricca Chemical Company
63516
500 mL Natural Poly Bottle
Each for $107.97
Only null left
 
635-32
View Documents
Ricca Chemical Company
63532
1 L Natural Poly Bottle
Each for $159.34
Only null left
 
635-1
View Documents
Ricca Chemical Company
RI6351
4 L Natural Poly Bottle
Each for $323.84
Only null left
 
R063500020F
View Documents
Ricca Chemical Company
R063500020F
20 L Cubitainer
Each for $498.25
Only null left
 
Description

Description

Specifications

Chemical Identifiers

1336-21-6
35.05
VHUUQVKOLVNVRT-UHFFFAOYSA-N
14923
amine hydrate
H5NO
MFCD00066650
ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh
CHEBI:18219
N.O

Specifications

1336-21-6,7732-18-5
MFCD00066650
500 mL
N.O
35.05
CHEBI:18219
Laboratory
Ammonium Hydroxide 20%, Ricca Chemical
H5NO
ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh
VHUUQVKOLVNVRT-UHFFFAOYSA-N
amine hydrate
14923
19.3 to 20.7%
Natural Poly Bottle
Videos
Safety and Handling

Safety and Handling

Recommended Storage : 15-30°C

SDS
missing translation for 'documents'

missing translation for 'documents'