missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NorthernMax™ High Stringency Wash Buffer DFS Item

The equivalent of 0.1X SSC or SSPE

Supplier:  Invitrogen™ AM8674

Numéro de catalogue. AM8674


Il en reste null
Découvrez plus d'offres spéciales

Description

Description

  • Functionally tested by northern analysis and is certified RNase-free
  • Same component included in the NorthernMax Ultrasensitive Northern Blotting Kit but in a larger size
  • Ready to use Comprehensive instructions are included

DNA and RNA Purification and Analysis, Northern Blotting, Nucleic Acid Gel Electrophoresis and Blotting

Order Info

Shipping Condition: Room Temperature

Spécifications

Spécifications

Store at room temperature.
Northern Analysis Wash Buffer
Room Temperature
Wash Buffer
1 L
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
FDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.