missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NorthernMax™-Gly Gel Prep/Running Buffer (10X) DFS Item

For the preparation and running of agarose gels

Supplier:  Invitrogen™ AM8678

Numéro de catalogue. AM8678

Il en reste null
Découvrez plus d'offres spéciales

Description

Description

  • For use with RNA denatured by glyoxal treatment
  • Same component included in the NorthernMax Ultrasensitive Northern Blotting Kits but in a larger size
  • These reagents are supplied ready to use and are certified RNase-free
  • Comprehensive instructions are included

Agarose Gel Electrophoresis, DNA and RNA Purification and Analysis, Northern Blotting, Nucleic Acid Gel Electrophoresis and Blotting

Order Info

Shipping Condition: Room Temperature

Spécifications

Spécifications

Store at 4°C.
Northern Analysis Gel Prep/Running Buffer
Room Temperature
10X
1 L
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
FDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.