Learn More
ADH7 Polyclonal Antibody
ADH7 Polyclonal Antibody
Supplier:  Thermo Fisher Scientific PA5104370
Numéro de catalogue. PIPA5104370
Il en reste null
Découvrez plus d'offres spéciales
Description
Description
ADH7 Polyclonal Antibody for Western Blot, ICC/IF
Spécifications
Spécifications
ADH7
Polyclonal
Unconjugated
ADH7
Adh3; Adh-3; Adh-3e; Adh3-e; Adh-3t; Adh3-t; Adh4; Adh7; ADH-C2; Adt-1; AI325182; alcohol dehydrogenase 3 complex; alcohol dehydrogenase 3 temporal; alcohol dehydrogenase 4; Alcohol dehydrogenase 7; alcohol dehydrogenase 7 (class IV), mu or sigma polypeptide; alcohol dehydrogenase class 4 mu/sigma chain; Alcohol dehydrogenase class IV mu/sigma chain; alcohol dehydrogenase VII; alcohol dehydrogenase-7; All-trans-retinol dehydrogenase [NAD(+)] ADH7; class IV alcohol dehydrogenase, mu or sigma subunit; class IV sigma-1 alcohol dehydrogenase; class IV sigmasigma alcohol dehydrogenase; gastric alcohol dehydrogenase; IV ADH; Omega-hydroxydecanoate dehydrogenase ADH7; ret; Retinol dehydrogenase
Rabbit
Affinity Chromatography
RUO
131, 171178
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P40394, P41682
ADH7
A synthesized peptide derived from human ADH7(Accession P40394), corresponding to amino acid residues D235-E285.
100 μL
Primary
Human, Rat
Antibody
IgG
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS