missing translation for 'onlineSavingsMsg'
Learn More

Acetonitrile, Reagent, ACS, 99.5%, Spectrum™ Chemical

A1030, 75-05-8, CH3CN

Supplier:  Spectrum Chemical Mfg Cor A10301LTGL

 Afficher plus de versions de ce produit

Numéro de catalogue. 18600296


Il en reste null
Découvrez plus d'offres spéciales

Description

Description

Spectrum™ Chemical Acetonitrile, Reagent, ACS is a colorless liquid with low viscosity and low chemical reactivity and is used in chemical labs as a polar aprotic solvent in organic synthesis. Acetonitrile is a medium-polarity solvent with a convenient liquid range that is miscible with water. As an ACS grade Reagent, Spectrum Chemical manufactured acetonitrile is used as the quality standard against which other substances are graded and has met the toughest regulatory standards for quality and purity.

Spécifications

Spécifications

Acetonitrile
100%
1 L
0.005%
C2H3N
UN1648
WEVYAHXRMPXWCK-UHFFFAOYSA-N
acetonitrile
99.5%
75-05-8
Clear
Amber Glass Bottle
8 μq/g
0.6 μeq/g
0.3%
CC#N
41.05
Reagent

Identifiants chimiques

75-05-8
41.05
acetonitrile
C2H3N
WEVYAHXRMPXWCK-UHFFFAOYSA-N
CC#N
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
FDS
Documents

Documents

Promotions

Promotions