missing translation for 'onlineSavingsMsg'
Learn More

Acetonitrile, Reagent, ACS, 99.5%, Spectrum™ Chemical

Catalog No. p-7156905
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
4 L
6 x 500 mL
20 L
4 x 4 L
6 x 1 L
Packaging:
Amber Glass Bottle
Steel Can
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18600290 100 mL Amber Glass Bottle
18600291 500 mL Amber Glass Bottle
18600296 1 L Amber Glass Bottle
18600292 6 x 500 mL Amber Glass Bottle
18600293 4 L Amber Glass Bottle
18600297 6 x 1 L Amber Glass Bottle
18600294 4 x 4 L Amber Glass Bottle
18600295 20 L Steel Can
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 18600290 Supplier Spectrum Chemical Mfg Cor Supplier No. A1030100ML
Only null left

A1030, 75-05-8, CH3CN

Spectrum™ Chemical Acetonitrile, Reagent, ACS is a colorless liquid with low viscosity and low chemical reactivity and is used in chemical labs as a polar aprotic solvent in organic synthesis. Acetonitrile is a medium-polarity solvent with a convenient liquid range that is miscible with water. As an ACS grade Reagent, Spectrum Chemical manufactured acetonitrile is used as the quality standard against which other substances are graded and has met the toughest regulatory standards for quality and purity.

Chemical Identifiers

CAS 75-05-8
Molecular Formula C2H3N
Molecular Weight (g/mol) 41.05
InChI Key WEVYAHXRMPXWCK-UHFFFAOYSA-N
IUPAC Name acetonitrile
SMILES CC#N

Specifications

CAS Max % 100%
Appearance Clear
Quantity 100 mL
Packaging Amber Glass Bottle
Residue after Evaporation 0.005%
Titratable Acid 8 μq/g
Titratable Base 0.6 μeq/g
UN Number UN1648
Water 0.3%
Percent Purity 99.5%
Grade Reagent
Chemical Name or Material Acetonitrile
Show More Show Less