missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetate Buffer, pH 3.5 For USP Heavy Metals Test, Reagents
Acetate Buffer, pH 3.5 For USP Heavy Metals Test, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Labchem Inc CA102300250A
Spécifications
| Acetate Buffer, pH 3.5 For USP Heavy Metals Test | |
| 118°C | |
| 3.5 | |
| 250 mL | |
| Poly Bottle | |
| Non-sterile |
| 7732-18-5,631-61-8,631-61-8 | |
| Colorless | |
| Liquid | |
| Buffer | |
| 1.0737 | |
| Reagent |