missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ T4 (Total) (Human), HRP, ELISA Kit
For quantitative measurement of Mouse Adiponectin/Acrp30 protein in serum and tissue culture medium
Supplier: Abnova™ KA0200
Comprend:
- Sheep anti-T4 coated microtiter wells 96 wells
- T4 Reference Standards: 0, 2, 5, 10, 15, and 25 ug/dl, ready to use 1 set, 1ml/vial
- Enzyme Conjugate Reagent Concentrate (11x) 1.3mL
- Enzyme Conjugate Diluent 13mL
- TMB Reagent (One-Step) 11mL
- Stop Solution (1N HCl) 11mL
Spécifications
| 0 to 25μg/dL | |
| Human | |
| 0.4 μg/dL | |
| 1 Kit | |
| Serum | |
| Store the kit at 4°C | |
| T4 (Total) (Human) ELISA Kit |
| HRP | |
| T4 (Total) (Human) ELISA Kit is a competitive enzyme immunoassay for the quantitative measurement of total thyroxine. | |
| Colorimetric | |
| For research use only (RUO) | |
| 25 μL | |
| Quantification |