missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ABCA13 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5149830
Description
ABCA13 Polyclonal Antibody for Western Blot, ICC/IF, IHC
In human, the ATP-binding cassette (ABC) family of transmembrane transporters has at least 48 genes and 7 gene subfamilies. This gene is a member of ABC gene subfamily A (ABCA). Genes within the ABCA family typically encode several thousand amino acids. Like other ABC transmembrane transporter proteins, this protein has 12 or more transmembrane alpha-helix domains that likely arrange to form a single central chamber with multiple substrate binding sites. It is also predicted to have two large extracellular domains and two nucleotide binding domains as is typical for ABCA proteins. Alternative splice variants have been described but their biological validity has not been demonstrated.[provided by RefSeq, Mar 2009].Spécifications
| ABCA13 | |
| Polyclonal | |
| Unconjugated | |
| ABCA13 | |
| ABCA13; ATP binding cassette subfamily A member 13; ATP binding cassette transporter A13; ATP-binding cassette sub-family A member 13; ATP-binding cassette sub-family A member 13 variant 2; ATP-binding cassette sub-family A member 13 variant 3; ATP-binding cassette, sub-family A (ABC1), member 13 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 268379 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry, Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q5SSE9 | |
| ABCA13 | |
| Recombinant ATP Binding Cassette Transporter A13 (ABCA13) (Met4692-Trp4931). | |
| 200 μL | |
| Primary | |
| Mouse | |
| Antibody |