missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BTX™ HT 100 Plate Handler, Manual Track Switching
Designed to work with 96 well plates. BTX™ Harvard Apparatus 96 Well Plate Handler helps to create a safe and reliable working area for processing electroporation plates.
Supplier: BTX™ 450400
Description
- Quickly optimize for better gene delivery
- Designed to work with 96 well plates
- Pulse switching technology integrated into package and gold plated contacts to mate with electroporation plate
- With an adapter to accommodate 25 well plates as well with our ECM 630 generator
- Prosess up to 96 wells in seconds
Spécifications
| 25-well, 96-well | |
| ECM630, ECM830, Gemini X2 electroporation systems | |
| Electroporation accessory | |
| HT-100 Microplate Handler | |
| Pin | |
| 1 to 99 | |
| 5°C to 40°C | |
| 20 to 80% | |
| Banana Cables | |
| Exponential Decay Wave, Square Wave |
| High-Throughput Electroporation | |
| BTX Electroporation | |
| 3000 VDC, 250 VAC | |
| LCD | |
| Manual track switching, Connectors for standard pulse generators | |
| 10 μs to 10 s | |
| 1 Ea. | |
| 2 mm, 4 mm | |
| Digital User | |
| Yes |