missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals 5-Methyl-DL-tryptophan, 98%
CAS: 951-55-3 | C12H14N2O2 | 218.26 g/mol
Supplier: Thermo Scientific Chemicals 127972500
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spécifications
| 5-Methyl-DL-tryptophan | |
| 280.0°C to 282.0°C | |
| Authentic | |
| Glass bottle | |
| MFCD00005652 | |
| 22,II,471 | |
| HUNCSWANZMJLPM-UHFFFAOYSA-N | |
| 2-amino-3-(5-methyl-1H-indol-3-yl)propanoic acid | |
| 92852 | |
| 218.25 | |
| Powder |
| 951-55-3 | |
| White | |
| 98% | |
| C12H14N2O2 | |
| 250 mg | |
| 5-methyl-dl-tryptophan, 5-methyltryptophan, 2-amino-3-5-methyl-1h-indol-3-yl propanoic acid, tryptophan, 5-methyl, dl-5-methyltryptophan, 5-methyl-tryptophan, 2-amino-3-5-methylindol-3-yl propanoic acid, 5-methyl tryptophan, dl, 5-methyltryptophan #, pubchem20652 | |
| CC1=CC=C2NC=C(CC(N)C(O)=O)C2=C1 | |
| 218.26 | |
| CHEBI:52524 | |
| 98% |
Identifiants chimiques
| 951-55-3 | |
| 218.26 | |
| HUNCSWANZMJLPM-UHFFFAOYSA-N | |
| 92852 | |
| 2-amino-3-(5-methyl-1H-indol-3-yl)propanoic acid |
| C12H14N2O2 | |
| MFCD00005652 | |
| 5-methyl-dl-tryptophan, 5-methyltryptophan, 2-amino-3-5-methyl-1h-indol-3-yl propanoic acid, tryptophan, 5-methyl, dl-5-methyltryptophan, 5-methyl-tryptophan, 2-amino-3-5-methylindol-3-yl propanoic acid, 5-methyl tryptophan, dl, 5-methyltryptophan #, pubchem20652 | |
| CHEBI:52524 | |
| CC1=CC=C2NC=C(CC(N)C(O)=O)C2=C1 |
Sécurité et manipulation
missing translation for 'einecsNumber' : 213-453-6
RUO – Research Use Only