missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 5-Methyl-DL-tryptophan, 98%

Catalog No. p-4509828
Change view
Click to view available options
Quantity:
250 mg
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC127972500 250 mg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC127972500 Supplier Thermo Scientific Chemicals Supplier No. 127972500
Only null left

CAS: 951-55-3 | C12H14N2O2 | 218.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 951-55-3
Molecular Formula C12H14N2O2
Molecular Weight (g/mol) 218.26
MDL Number MFCD00005652
InChI Key HUNCSWANZMJLPM-UHFFFAOYSA-N
Synonym 5-methyl-dl-tryptophan, 5-methyltryptophan, 2-amino-3-5-methyl-1h-indol-3-yl propanoic acid, tryptophan, 5-methyl, dl-5-methyltryptophan, 5-methyl-tryptophan, 2-amino-3-5-methylindol-3-yl propanoic acid, 5-methyl tryptophan, dl, 5-methyltryptophan #, pubchem20652
PubChem CID 92852
ChEBI CHEBI:52524
IUPAC Name 2-amino-3-(5-methyl-1H-indol-3-yl)propanoic acid
SMILES CC1=CC=C2NC=C(CC(N)C(O)=O)C2=C1

Specifications

Melting Point 280.0°C to 282.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 250 mg
Beilstein 22,II,471
Formula Weight 218.25
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 5-Methyl-DL-tryptophan
Show More Show Less
EINECSNumber 213-453-6

RUO – Research Use Only