missing translation for 'onlineSavingsMsg'
Learn More

Sigma Aldrich 5-Hydroxy-1,4-naphthoquinone

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

$122.10 - $122.10

Spécifications

CAS 481-39-0
Melting Point 161°C to 163°C (lit.)
Molecular Formula C10H6O3
Linear Formula C10H6O3
MDL Number MFCD00001684
voir les spécifications

Produits 1
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantité  
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantité  
H470031G
Afficher les documents
Sigma Aldrich
H470031G
1 g
chaque for $122.10
Il en reste null
 
Description

Description

5-Hydroxy-1,4-naphthoquinone
Spécifications

Spécifications

481-39-0
C10H6O3
MFCD00001684
174.15
161°C to 163°C (lit.)
C10H6O3
Juglone
97%
Videos
FDS
missing translation for 'documents'

missing translation for 'documents'